missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-methylpyridine, 97%, Thermo Scientific Chemicals

Product Code. 11431830
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431830 1 g 1g
15403717 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11431830 Supplier Thermo Scientific Alfa Aesar Supplier No. H27540.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3430-10-2
MDL Number MFCD03788195

Specifications

Melting Point 114°C to 119°C
Odor Characteristic
Molecular Formula C20H40
Quantity 1 g
UN Number UN2811
Synonym 1-eicosene, 1-icosene, eicosene, cetyl ethylene, .alpha.-eicosene, unii-6bhh7zd637, arachidene, arachydene, cosen, alpha-eicosene
InChI Key VAMFXQBUQXONLZ-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCCC=C
IUPAC Name icos-1-ene
Molecular Weight (g/mol) 280.54
PubChem CID 18936
Formula Weight 108.14
Percent Purity 97%
Chemical Name or Material 3-Amino-2-methylpyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.