missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-methylpyridine, 97%, Thermo Scientific™

Product Code. 11996381 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11996381 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11996381 Supplier Thermo Scientific Acros Supplier No. 437630010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Specifications

CAS 3430-10-2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 116°C to 118°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C6H8N2
Quantity 1 g
Synonym 1-eicosene, 1-icosene, eicosene, cetyl ethylene, .alpha.-eicosene, unii-6bhh7zd637, arachidene, arachydene, cosen, alpha-eicosene
InChI Key ZSFPJJJRNUZCEV-UHFFFAOYSA-N
SMILES CC1=C(N)C=CC=N1
IUPAC Name icos-1-ene
Molecular Weight (g/mol) 108.14
PubChem CID 18936
Formula Weight 108.14
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 3-Amino-2-methylpyridine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-2-methylpyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.