missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 3-Amino-3-Deoxydigoxigenin Hemisuccinamide, Succinimidyl Ester

Product Code. 11564037
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11564037 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11564037 Supplier Invitrogen™ Supplier No. A2952
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3-amino-3-deoxydigoxigenin hemisuccinamide, succinimidyl ester

The amine-reactive version of digoxigenin (3-amino-3-deoxygigoxigenin hemisuccinamide, SE) can be used to attach this important hapten to biomolecules of interest for subsequent detection with an anti-digoxigenin antibody.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type 3-Amino-3-Deoxydigoxigenin Hemisuccinamide Succinimidyl Ester
Content And Storage Store in freezer (-5 to -30°C).
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Digoxigenin
Label Type Biotin and Other Haptens

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.