missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-methyloxetane, 95%

Product Code. 15444048
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15444048 500 mg 500mg
15434048 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444048 Supplier Thermo Scientific Alfa Aesar Supplier No. H57588.ME
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 874473-14-0 | C4H9NO | 87.122 g/mol

It is used as an OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an OLED intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874473-14-0
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD11111713
InChI Key NQVWMPOQWBDSAI-UHFFFAOYSA-N
Synonym 3-methyl-3-oxetanamine, 3-amino-3-methyloxetane, 3-oxetanamine,3-methyl, 3-oxetanamine, 3-methyl, pubchem14416, pubchem23160, acmc-209qnl, 3-methyl 3-oxetanamine, 3-methyl-3-oxetaneamine, 3-amino-3-methyl-oxetane
PubChem CID 46835725
IUPAC Name 3-methyloxetan-3-amine
SMILES CC1(COC1)N

Specifications

Density 0.99 g/mL
Boiling Point 102°C
Quantity 500 mg
Solubility Information Slightly soluble in water.
Formula Weight 87.12
Percent Purity 95%
Physical Form Crystalline Solid
Chemical Name or Material 3-Amino-3-methyloxetane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.