missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-phenyl-1-propanol, 94%

Product Code. 10199512 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10199512 500 mg Glass bottle 500mg
11945281 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10199512 Supplier Thermo Scientific Acros Supplier No. 292935000
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 14593-04-5 | C9H13NO | 151.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14593-04-5
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00801124,MFCD01311768,MFCD01311791
InChI Key SEQXIQNPMQTBGN-UHFFFAOYNA-N
Synonym 3-amino-3-phenyl-1-propanol, dl-beta-phenylalaninol, 3-amino-3-phenyl-propan-1-ol, 3-phenyl-dl-beta-alaninol, 1-propanol, 3-amino-3-phenyl, benzenepropanol, gamma-amino, 1-amino-3-hydroxyprop-1-yl benzene, dl--phenylalaninol, 3-amino-3-phenylpropanol, acmc-1c4c0
PubChem CID 203500
IUPAC Name 3-amino-3-phenylpropan-1-ol
SMILES NC(CCO)C1=CC=CC=C1

Specifications

Melting Point 70.0°C to 77.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Linear Formula C6H5CH(NH2)CH2CH2OH
Quantity 500 mg
Beilstein 13,IV,1919
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol, ethanol and chloroform
Formula Weight 151.21
Percent Purity 94%
Physical Form Needles
Chemical Name or Material 3-Amino-3-phenyl-1-propanol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-3-phenyl-1-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.