missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-4-[(1-benzyl-2-methoxy-2-oxoethyl)amino]-4-oxobutanoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
Specifications
Specifications
| CAS Min % | 97.0 |
| Assay Percent Range | 97% |
| Quantity | 100 g |
| IUPAC Name | 3-amino-4-[(1-methoxy-1-oxo-3-phenylpropan-2-yl)amino]-4-oxobutanoic acid |
| Formula Weight | 294.31 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-Amino-4-[(1-benzyl-2-methoxy-2-oxoethyl)amino]-4-oxobutanoic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction