missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4,5-dihydro-1-phenyl-1H-pyrazole, 98+%

Product Code. 11392526
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392526 10 g 10g
11382526 2 g 2g
11302536 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11392526 Supplier Thermo Scientific Alfa Aesar Supplier No. L06476.09
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3314-35-0 | C9H11N3 | 161.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3314-35-0
Molecular Formula C9H11N3
Molecular Weight (g/mol) 161.208
MDL Number MFCD00051730
InChI Key QENUTIJJGGTTPE-UHFFFAOYSA-N
Synonym 1-phenyl-4,5-dihydro-1h-pyrazol-3-amine, 2-pyrazoline, 3-amino-1-phenyl, 1-fenyl-3-aminopyrazolin, 3-amino-1-phenyl-2-pyrazoline, 1-phenyl-4,5-dihydro-1h-pyrazol-3-ylamine, 3-amino-4,5-dihydro-1-phenylpyrazole, pyrazolidine, 3-imino-1-phenyl, 4,5-dihydro-1-phenyl-1h-pyrazol-3-amine, 3-amino-1-fenyl-2-pyrazolin, 1-phenyl-3-amino-2-pyrazoline
PubChem CID 98736
IUPAC Name 2-phenyl-3,4-dihydropyrazol-5-amine
SMILES C1CN(N=C1N)C2=CC=CC=C2

Specifications

Melting Point 167°C to 170°C
Quantity 10 g
Beilstein 139162
Formula Weight 161.21
Percent Purity ≥98%
Chemical Name or Material 3-Amino-4,5-dihydro-1-phenyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.