missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-bromo-5-methylisoxazole, 97%

Product Code. 11430671
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430671 5 g 5g
11420671 1 g 1g
11440671 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430671 Supplier Thermo Scientific Alfa Aesar Supplier No. B20138.06
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 5819-40-9 | C4H5BrN2O | 177.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5819-40-9
Molecular Formula C4H5BrN2O
Molecular Weight (g/mol) 177.00
MDL Number MFCD00052553
InChI Key JEZOZNWEHSNXPQ-UHFFFAOYSA-N
Synonym 3-amino-4-bromo-5-methylisoxazole, 4-bromo-5-methylisoxazol-3-amine, 3-isoxazolamine, 4-bromo-5-methyl, 4-bromo-5-methylisoxazole-3-ylamine, acmc-20ana7, 4-bromo-5-methyl-3-isoxazolamine, 4-bromo-5-methyl-isoxazol-3-ylamine, 4-bromanyl-5-methyl-1,2-oxazol-3-amine, 3-amino-4-bromo-5-methyl isoxazole
PubChem CID 2774464
IUPAC Name 4-bromo-5-methyl-1,2-oxazol-3-amine
SMILES CC1=C(Br)C(N)=NO1

Specifications

Melting Point 73°C to 75°C
Quantity 5 g
Formula Weight 177.02
Percent Purity 97%
Chemical Name or Material 3-Amino-4-bromo-5-methylisoxazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.