missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-carbethoxypyrazole, 99%

Product Code. 10050580 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10050580 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10050580 Supplier Thermo Scientific Acros Supplier No. 103310050
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6994-25-8 | C6H9N3O2 | 155.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6994-25-8
CAS Min % 98.5
CAS Max % 100.0
Melting Point 103.0°C to 107.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C6H9N3O2
MDL Number MFCD00005238
Quantity 5 g
Synonym ethyl 3-amino-1h-pyrazole-4-carboxylate, ethyl 3-amino-4-pyrazolecarboxylate, 3-amino-4-carbethoxypyrazole, 5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, 3-amino-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 3-aminopyrazole-4-carboxylate, 3-amino-4-ethoxycarbonylpyrazole, unii-9gy5ha1gaa, 3-amino-4-carbethoxypyrazol, 1h-pyrazole-4-carboxylic acid, 3-amino-, ethyl ester
InChI Key YPXGHKWOJXQLQU-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(NN=C1)N
IUPAC Name ethyl 5-amino-1H-pyrazole-4-carboxylate
Molecular Weight (g/mol) 155.16
PubChem CID 81472
Formula Weight 155.16
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-carbethoxypyrazole, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.