missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-ethyl-1H-pyrazole, 98%

Product Code. 15419768
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15419768 1 g 1g
15439768 250 mg 250mg
15429768 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15419768 Supplier Thermo Scientific Alfa Aesar Supplier No. H63199.03
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 43024-15-3 | C5H9N3 | 111.15 g/mol

3-Amino-4-ethyl-1H-pyrazole is employed as a intermediate for chemical research and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-4-ethyl-1H-pyrazole is employed as a intermediate for chemical research and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43024-15-3
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.15
MDL Number MFCD06797570
InChI Key RDCODVKTTJWFAR-UHFFFAOYSA-N
Synonym 4-ethyl-1h-pyrazol-3-amine, 3-amino-4-ethylpyrazole, 3-amino-4-ethyl-1h-pyrazole, 4-ethyl-2h-pyrazol-3-amine, 3-amino-4-ethyl pyrazole, 4-ethyl-1h-pyrazol-3-ylamine, 1h-pyrazol-3-amine, 4-ethyl
PubChem CID 11789157
IUPAC Name 4-ethyl-1H-pyrazol-5-amine
SMILES CCC1=C(N)NN=C1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 111.15
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Amino-4-ethyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.