missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-iodo-1H-indazole, 95%, Thermo Scientific™

Product Code. 10199359 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
10199359 5 g 5g
10311730 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 10199359

Brand: Thermo Scientific Acros 435750050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 599191-73-8
Molecular Formula C7H6IN3
Molecular Weight (g/mol) 259.05
MDL Number MFCD03426697
InChI Key OEQIVIYSUJXCFG-UHFFFAOYSA-N
Synonym 4-iodo-1h-indazol-3-ylamine, 3-amino-4-iodo-1h-indazole, 1h-indazol-3-amine, 4-iodo, 3-amino-4-iodoindazole, 4-iodo-1h-indazol-amine, ksc268o0n, 4-iodo-1h-indazole-3-amine, 4-iodo-1h-indazole-3-ylamine, 3-amino-4-iodo-indazole
PubChem CID 4057787
IUPAC Name 4-iodo-1H-indazol-3-amine
SMILES NC1=NNC2=CC=CC(I)=C12

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 150.0°C to 152.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 259.05
Percent Purity 95%
Physical Form Crystalline Solid
Chemical Name or Material 3-Amino-4-iodo-1H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-4-iodo-1H-indazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.