missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-cyclobutyl-1H-pyrazole, 97%

Product Code. 11479570
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11479570 250 mg 250mg
11489570 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11479570

Brand: Thermo Scientific Alfa Aesar H50880.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 326827-21-8 | C7H11N3 | 137.19 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 326827-21-8
Molecular Formula C7H11N3
Molecular Weight (g/mol) 137.19
MDL Number MFCD08061045,MFCD11187387
InChI Key DELFRVWPWUEOHU-UHFFFAOYSA-N
Synonym 3-amino-5-cyclobutyl-1h-pyrazole, 3-cyclobutyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-cyclobutyl, 5-cyclobutyl-1h-pyrazol-3-ylamine, 5-cyclobutyl-2h-pyrazol-3-amine, 5-cyclobutyl-2h-pyrazol-3-ylamine, 3-amino-5-cyclobutyl-2h-pyrazole, acmc-20agky, 5-amino-3-cyclobutylpyrazole
PubChem CID 22176774
IUPAC Name 5-cyclobutyl-1H-pyrazol-3-amine
SMILES NC1=NNC(=C1)C1CCC1

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 137.19
Percent Purity 97%
Chemical Name or Material 3-Amino-5-cyclobutyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.