missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-cyclobutyl-1H-pyrazole, 97%

Product Code. 11489570
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489570 1 g 1g
11479570 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489570 Supplier Thermo Scientific Alfa Aesar Supplier No. H50880.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 326827-21-8 | C7H11N3 | 137.19 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 326827-21-8
Molecular Formula C7H11N3
Molecular Weight (g/mol) 137.19
MDL Number MFCD08061045,MFCD11187387
InChI Key DELFRVWPWUEOHU-UHFFFAOYSA-N
Synonym 3-amino-5-cyclobutyl-1h-pyrazole, 3-cyclobutyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-cyclobutyl, 5-cyclobutyl-1h-pyrazol-3-ylamine, 5-cyclobutyl-2h-pyrazol-3-amine, 5-cyclobutyl-2h-pyrazol-3-ylamine, 3-amino-5-cyclobutyl-2h-pyrazole, acmc-20agky, 5-amino-3-cyclobutylpyrazole
PubChem CID 22176774
IUPAC Name 5-cyclobutyl-1H-pyrazol-3-amine
SMILES NC1=NNC(=C1)C1CCC1

Specifica

Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 137.19
Percent Purity 97%
Chemical Name or Material 3-Amino-5-cyclobutyl-1H-pyrazole

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.