missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-ethyl-1H-pyrazole, 97%, Thermo Scientific™

Product Code. 10037839
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10037839 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10037839 Supplier Thermo Scientific Maybridge Supplier No. MAY00221DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1904-24-1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C5H9N3
MDL Number MFCD08060982
Quantity 1 g
Synonym 3-ethyl-1h-pyrazol-5-amine, 5-amino-3-ethyl-1h-pyrazole, 3-amino-5-ethylpyrazole, 3-amino-5-ethyl-1h-pyrazole, 5-ethyl-2h-pyrazol-3-ylamine, 1h-pyrazol-3-amine, 5-ethyl, 5-ethyl-2h-pyrazol-3-amine, ethyl-5-aminopyrazole, 3-amino-5-ethypyrazole, 1h-pyrazol-5-amine, 3-ethyl
InChI Key AXDGPQLEVYSXNL-UHFFFAOYSA-N
SMILES CCC1=CC(N)=NN1
IUPAC Name 5-ethyl-1H-pyrazol-3-amine
Molecular Weight (g/mol) 111.15
PubChem CID 21991095
Formula Weight 111.15
Percent Purity 97%
Chemical Name or Material 3-Amino-5-ethyl-1H-pyrazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.