missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-(methoxycarbonyl)benzeneboronic acid hydrochloride, 97%, Thermo Scientific™

Product Code. 15440196
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440196 250 mg 250mg
15430196 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440196 Supplier Thermo Scientific Alfa Aesar Supplier No. H52904.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380430-56-8
Molecular Formula C8H11BClNO4
Molecular Weight (g/mol) 231.439
MDL Number MFCD04971992
InChI Key NOMZZWSCDMMVAF-UHFFFAOYSA-N
Synonym 3-amino-5-methoxycarbonylphenylboronic acid, hcl, 3-amino-5-methoxycarbonyl phenyl boronic acid hydrochloride, 3-amino-5-methoxycarbonylphenyl boronic acid hydrochloride, 3-amino-5-methoxycarbonyl phenylboronic acid hydrochloride, 3-amino-5-methoxycarbonylphenylboronic acid hydrochloride, 3-amino-5-methoxycarbonylphenylboronic acid hcl, benzoicacid, 3-amino-5-borono-, 1-methyl ester, 3-amino-5-methoxycarbonyl phenylboronic acid hcl, 3-azanyl-5-methoxycarbonyl-phenyl boronic acid hydrochloride, 3-amino-5-methoxycarbonyl phenyl boronic acid-hydrogen chloride 1/1
PubChem CID 16427084
IUPAC Name (3-amino-5-methoxycarbonylphenyl)boronic acid;hydrochloride
SMILES B(C1=CC(=CC(=C1)N)C(=O)OC)(O)O.Cl

Specifications

Melting Point 220°C to 226°C
Quantity 250 mg
Formula Weight 231.44
Percent Purity 97%
Chemical Name or Material 3-Amino-5-(methoxycarbonyl)benzeneboronic acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.