missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-(tert-butyl)thiophene-2-carboxamide, 97%, Thermo Scientific™

Product Code. p-2698355
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10572012 1 g 1g
10364522 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10572012 Supplier Thermo Scientific Maybridge Supplier No. GK00992DA
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175137-04-9
Molecular Formula C9H14N2OS
Molecular Weight (g/mol) 198.28
MDL Number MFCD00068165
InChI Key MKHZJBZWKNDDCN-UHFFFAOYSA-N
Synonym 3-amino-5-tert-butyl thiophene-2-carboxamide, 3-amino-5-tert-butyl thiophene-2-carboxamid, 2-thiophenecarboxamide,3-amino-5-1,1-dimethylethyl, 3-amino-5-tert-butyl thiophene-2-carboximidic acid, pubchem13396, maybridge1_003717, 3-amino-5-t-butyl thiophene-2-carboxamide, 3-amino-5-tert-butyl-2-thiophenecarboxamide, 5-tert-butyl-3-aminothiophene-2-carboxamide, 3-amino-5-tert-buty thiophene-2-carboxamide
PubChem CID 2735293
IUPAC Name 3-amino-5-tert-butylthiophene-2-carboxamide
SMILES CC(C)(C)C1=CC(N)=C(S1)C(N)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 198.29
Percent Purity 97%
Chemical Name or Material 3-Amino-5-(tert-butyl)thiophene-2-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.