missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-tert-butylisoxazole, 97%

Product Code. 11498947
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498947 25 g 25g
11488947 5 g 5g
11408957 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498947 Supplier Thermo Scientific Alfa Aesar Supplier No. B20567.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 55809-36-4 | C7H12N2O | 140.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55809-36-4
Molecular Formula C7H12N2O
Molecular Weight (g/mol) 140.186
MDL Number MFCD00055620
InChI Key GGXGVZJHUKEJHO-UHFFFAOYSA-N
Synonym 3-amino-5-tert-butylisoxazole, 5-tert-butylisoxazol-3-amine, 3-amino-5-t-butylisoxazole, 5-tert-butyl isoxazol-3-amine, 3-isoxazolamine, 5-1,1-dimethylethyl, 3-amino-5-tertbutylisoxazole, 5-tert-butyl-isoxazol-3-ylamine, 3-amino-5-tert-butyl isoxazole, 5-tert-butyl isoxazole-3-ylamine
PubChem CID 171473
IUPAC Name 5-tert-butyl-1,2-oxazol-3-amine
SMILES CC(C)(C)C1=CC(=NO1)N

Specifications

Melting Point 110°C to 113°C
Quantity 25 g
Beilstein 1100990
Formula Weight 140.19
Percent Purity 97%
Chemical Name or Material 3-Amino-5-tert-butylisoxazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.