missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-(trifluoromethyl)-1H-indazole, 97%, Thermo Scientific™

Product Code. 15435787
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435787 250 mg 250mg
15425787 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435787 Supplier Thermo Scientific Alfa Aesar Supplier No. H32995.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2250-53-5
Molecular Formula C8H6F3N3
Molecular Weight (g/mol) 201.152
MDL Number MFCD01719740
InChI Key ZXZTZRVCWTWKCH-UHFFFAOYSA-N
Synonym 5-trifluoromethyl-1h-indazol-3-amine, 3-amino-5-trifluoromethyl-1h-indazole, 1h-indazol-3-amine, 5-trifluoromethyl, 1h-indazol-3-amine, 5-trifluoromethyl-9ci, 1h-indazole, 3-amino-5-trifluoromethyl, 5-trifluoromethyl-1h-indazole-3-ylamine, 3-amino-5-trifluoromethylindazole, 3-amino-5trifluoromethyl-indazole, 5-trifluoromethyl-1h-indazol-3-ylamine
PubChem CID 16736
IUPAC Name 5-(trifluoromethyl)-1H-indazol-3-amine
SMILES C1=CC2=C(C=C1C(F)(F)F)C(=NN2)N

Specifications

Melting Point 133°C to 136°C
Odor Odorless
Quantity 250 mg
UN Number UN2811
Formula Weight 201.15
Percent Purity 97%
Chemical Name or Material 3-Amino-5-(trifluoromethyl)-1H-indazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.