missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-6-bromo-1H-indazole, 97%

Product Code. 15437007
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437007 10 g 10g
15427007 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437007 Supplier Thermo Scientific Alfa Aesar Supplier No. H34095.09
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 404827-77-6 | C7H6BrN3 | 212.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 404827-77-6
Molecular Formula C7H6BrN3
Molecular Weight (g/mol) 212.05
MDL Number MFCD05665872
InChI Key WLDHNAMVDBASAW-UHFFFAOYSA-N
PubChem CID 2786631
IUPAC Name 6-bromo-1H-indazol-3-amine
SMILES C1=CC2=C(C=C1Br)NN=C2N

Specifications

Melting Point 227°C to 230°C
Quantity 10 g
Synonym 3-amino-6-bromo-1h-indazole, 6-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 6-bromo, 6-bromo-1h-indazole-3-amine, pubchem17937, acmc-209jef, 6-bromo-3-amino-1h-indazole, 1h-indazol-3-amine,6-bromo, 3-amino-6-bromoindazole, 6-bromo-1h-indazole-3-ylamine
Formula Weight 212.05
Percent Purity 97%
Chemical Name or Material 3-Amino-6-bromo-1H-indazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.