missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-N,N-dimethyl-4-nitroaniline, 97%, Thermo Scientific™

Product Code. 10459281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
Product Code. Quantity Packaging unitSize
10459281 5 g Glass bottle 5g
1 options
This item is not returnable. View return policy

Product Code. 10459281

Brand: Thermo Scientific Acros 326220050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 2069-71-8
CAS Min % 96.0
CAS Max % 100.0
Melting Point 134°C to 139°C
Color Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H11N3O2
MDL Number MFCD01320677
Quantity 5 g
Beilstein 13, IV, 97
Synonym 3-amino-n,n-dimethyl-4-nitroaniline, n1,n1-dimethyl-4-nitrobenzene-1,3-diamine, 3-amino n,n-dimethyl 4-nitro aniline, 5-dimethylamino-2-nitroaniline, n1,n1-dimethyl-4-nitro-benzene-1,3-diamine, n1,n1-dimethyl-4-nitro-m-phenylendiamin, 1,3-benzenediamine,n1,n1-dimethyl-4-nitro, 3-amino-4-nitro-n,n-dimethylaniline, 3-amino-4-nitrodimethylaminobenzene, 3-amino-4-nitrophenyl dimethylamine
InChI Key WJTOMXLUNDWLCY-UHFFFAOYSA-N
SMILES CN(C)C1=CC=C(C(N)=C1)[N+]([O-])=O
IUPAC Name 1-N,1-N-dimethyl-4-nitrobenzene-1,3-diamine
Molecular Weight (g/mol) 181.20
PubChem CID 2733451
Formula Weight 181.19
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Amino-N, N-dimethyl-4-nitroaniline, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-N,N-dimethyl-4-nitroaniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.