missing translation for 'onlineSavingsMsg'
Learn More

3'-Aminoacetophenone, 97%, ACROS Organics™

Product Code. 10473775 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10473775 100 g Glass bottle 100g
10502761 5 g Glass bottle 5g
10104080 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10473775 Supplier Thermo Scientific Acros Supplier No. 160771000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 99-03-6
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00007796
InChI Key CKQHAYFOPRIUOM-UHFFFAOYSA-N
Synonym 3'-aminoacetophenone, 1-3-aminophenyl ethanone, m-aminoacetophenone, 3-aminoacetophenone, 3-acetylaniline, m-acetylaniline, ethanone, 1-3-aminophenyl, 1-3-aminophenyl ethan-1-one, m-aminoacetylbenzene, acetophenone, 3'-amino
PubChem CID 7417
IUPAC Name 1-(3-aminophenyl)ethan-1-one
SMILES CC(=O)C1=CC=CC(N)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2NC6H4COCH3
Quantity 100 g
Beilstein 14, 45
Merck Index 14, 413
Formula Weight 135.17
Chemical Name or Material 3';-Aminoacetophenone, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.