missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzamide, 98%

Product Code. 11459712
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459712 25 g 25g
11449712 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459712 Supplier Thermo Scientific Alfa Aesar Supplier No. A10793.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3544-24-9 | C7H8N2O | 136.15 g/mol

3-Aminobenzamide is used as an endogenous poly-ADP-ribosyltransferase inhibitor. Through its effects on PARPs, 3-amino benzamide causes telomere shortening and stimulates angiogenesis. It displays neuroprotective, anti-necrotic effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminobenzamide is used as an endogenous poly-ADP-ribosyltransferase inhibitor. Through its effects on PARPs, 3-amino benzamide causes telomere shortening and stimulates angiogenesis. It displays neuroprotective, anti-necrotic effects.

Solubility
Soluble in dimethylformamide (∼30 mg/ml), dimethyl sulfoxide (∼30 mg/ml), phosphate buffered saline (pH7.2) (∼2 mg/ml), water (25 mg/ml), and ethanol (25 mg/ml).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3544-24-9
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00007989
InChI Key GSCPDZHWVNUUFI-UHFFFAOYSA-N
Synonym m-aminobenzamide, benzamide, 3-amino, 3-amino-benzamide, benzamide, m-amino, meta-aminobenzamide, 3-aba, 3-aminobenzimide, 3-amino benzamide, aniline-3-carboxamide, 3-aminobenzoic acid amide
PubChem CID 1645
ChEBI CHEBI:64042
IUPAC Name 3-aminobenzamide
SMILES NC(=O)C1=CC=CC(N)=C1

Specifications

Melting Point 111°C to 115°C
Quantity 25 g
Beilstein 2802373
Solubility Information Soluble in dimethylformamide (∽30mg/ml),dimethyl sulfoxide (∽30mg/ml),phosphate buffered saline (pH7.2) (∽2mg/ml),water (25mg/ml),and ethanol (25mg/ml).
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 3-Aminobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.