missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzeneboronic acid hydrochloride, 98%

Product Code. 15449357
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449357 5 g 5g
15439357 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449357 Supplier Thermo Scientific Alfa Aesar Supplier No. H52595.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 85006-23-1 | C6H9BClNO2 | 173.40 g/mol

3-Aminobenzeneboronic acid hydrochloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminobenzeneboronic acid hydrochloride is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85006-23-1
Molecular Formula C6H9BClNO2
Molecular Weight (g/mol) 173.40
MDL Number MFCD00191748
InChI Key QBMHZZHJIBUPOX-UHFFFAOYSA-N
Synonym 3-aminophenyl boronic acid hydrochloride, 3-aminophenylboronic acid hydrochloride, 3-aminophenylboronic acid hcl, m-aminophenylboronic acid hydrochloride, 3-aminophenylboronic acid, hcl, 3-aminobenzeneboronic acid hydrochloride, boronic acid, 3-aminophenyl-, hydrochloride, pubchem1751, pubchem20396, timtec-bb sbb003816
PubChem CID 2763241
IUPAC Name (3-aminophenyl)boronic acid;hydrochloride
SMILES Cl.NC1=CC=CC(=C1)B(O)O

Specifications

Melting Point >300°C
Quantity 5 g
Formula Weight 173.41
Percent Purity 98%
Chemical Name or Material 3-Aminobenzeneboronic acid hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.