missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzeneboronic acid pinacol ester, 97%

Product Code. 11452050
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452050 5 g 5g
11442050 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11452050 Supplier Thermo Scientific Alfa Aesar Supplier No. H28448.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 210907-84-9 | C12H18BNO2 | 219.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210907-84-9
Molecular Formula C12H18BNO2
Molecular Weight (g/mol) 219.09
MDL Number MFCD03453668
InChI Key YMXIIVIQLHYKOT-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-aminophenylboronic acid pinacol ester, benzenamine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-aminophenylboronic acid, pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-aminobenzeneboronic acid pinacol ester, 3-aminophenylboronic acid, pinacol cyclic ester, 3-aminophenylboronicacidpinacolester, apbe, amtb060
PubChem CID 2734655
IUPAC Name 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC(N)=C1

Specifications

Melting Point 87°C to 93°C
Quantity 5 g
Formula Weight 219.09
Percent Purity 97%
Chemical Name or Material 3-Aminobenzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.