missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzenesulfonamide, 97+%

Product Code. 11464907
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464907 5 g 5g
11474907 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464907 Supplier Thermo Scientific Alfa Aesar Supplier No. B21857.06
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 98-18-0 | C6H8N2O2S | 172.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-18-0
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00035781
InChI Key JPVKCHIPRSQDKL-UHFFFAOYSA-N
Synonym metanilamide, m-aminobenzenesulfonamide, benzenesulfonamide, 3-amino, m-sulfamoylaniline, 3-aminobenzene-1-sulfonamide, benzenesulfonamide, m-amino, m-aminobenzenesulphonamide, metaniilamide, 3-aminosulfonylaniline, 3-amino-benzenesulfonamide
PubChem CID 7377
IUPAC Name 3-aminobenzenesulfonamide
SMILES C1=CC(=CC(=C1)S(=O)(=O)N)N

Specifications

Melting Point 139°C to 142°C
Quantity 5 g
Formula Weight 172.21
Percent Purity ≥97%
Chemical Name or Material 3-Aminobenzenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.