missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzhydrazide, 97%, Thermo Scientific™

Product Code. 11470393
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470393 250 g 250g
11450393 10 g 10g
11460393 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470393 Supplier Thermo Scientific Alfa Aesar Supplier No. A11491.30
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14062-34-1
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.17
MDL Number MFCD00017067
InChI Key JSIVTKBYJLGBPY-UHFFFAOYSA-N
Synonym 3-aminobenzhydrazide, m-aminobenzohydrazide, 3-aminobenzoyl hydrazine, m-aminobenzhydrazide, 3-amino-benzoic acid hydrazide, m-amino benzhydrazide, benzoic acid, 3-amino-, hydrazide, m-aminobenzoic acid hydrazide, 3-???phenylamine, 3-aminobenzoylhydrazine
PubChem CID 84178
IUPAC Name 3-aminobenzohydrazide
SMILES NNC(=O)C1=CC=CC(N)=C1

Specifications

Melting Point 90°C to 94°C
Quantity 250 g
Beilstein 2090097
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material 3-Aminobenzhydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.