missing translation for 'onlineSavingsMsg'
Learn More

3-(Aminocarbonyl)-1H-indol-5-aminium chloride, Thermo Scientific™

Product Code. 10632653
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10632653 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10632653 Supplier Thermo Scientific Maybridge Supplier No. SP01297DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 306936-36-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H10ClN3O
Quantity 1 g
Synonym 5-amino-1h-indole-3-carboxamide hydrochloride, 3-aminocarbonyl-1h-indol-5-amine hydrochloride, 3-aminocarbonyl-1h-indol-5-aminium chloride, 5-aminoindole-3-carboxamide, chloride, 5-amino-1h-indole-3-carboxamide hydrochloride, tech, 5-amino-1h-indole-3-carboxamide-hydrogen chloride 1/1, 3-aminocarbonyl-1h-indol-5-amine hydrochloride 0.5 hydrate, 3-aminocarbonyl-1h-indol-5-amine hydrochloride hemihydrate
InChI Key GCGFMKAKROLCMQ-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C1N)C(=CN2)C(=O)N.Cl
IUPAC Name 5-amino-1H-indole-3-carboxamide;hydrochloride
Molecular Weight (g/mol) 211.649
PubChem CID 24229789
Formula Weight 220.66
Percent Purity 97%
Chemical Name or Material 3-(Aminocarbonyl)-1H-indol-5-Amine hydrochloride 0.5 hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.