missing translation for 'onlineSavingsMsg'
Learn More

3-Aminocrotononitrile, (E)+(Z), 96%

Product Code. 11337395
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
missing translation for 'unitSize':
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
11337395 500 g 500g
11327395 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11337395 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' L05316.36
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1118-61-2 | C4H6N2 | 82.11 g/mol

3-Aminocrotononitrile reacts with ferrocenyl-1,2-enones to form ferrocenyl pyridines. It undergoes diazotization coupling reaction with p-substituted anilines to give 2-arylhydrazone-3-ketimino-butyronitriles.3-Aminocrotononitrile, (E)+(Z), is used in reaction with aryldiazonium salts gave, instead of the expected triazene, 2-arylhydrazono-3-ketobutyronitrile by electrophilic attack at the ?-carbon and hydrolysis of the resulting imine. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Aminocrotononitrile was used as bisnucleophilic reagent which on cyclocondensation with hexafluoroacetone(ethoxycarbonylimine) forms bis(trifluoromethyl)pyrimidinones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminocrotononitrile reacts with ferrocenyl-1,2-enones to form ferrocenyl pyridines. It undergoes diazotization coupling reaction with p-substituted anilines to give 2-arylhydrazone-3-ketimino-butyronitriles.3-Aminocrotononitrile, (E)+(Z), is used in reaction with aryldiazonium salts gave, instead of the expected triazene, 2-arylhydrazono-3-ketobutyronitrile by electrophilic attack at the ɑ-carbon and hydrolysis of the resulting imine. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Aminocrotononitrile was used as bisnucleophilic reagent which on cyclocondensation with hexafluoroacetone(ethoxycarbonylimine) forms bis(trifluoromethyl)pyrimidinones.

Solubility
Very soluble in water, soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-61-2
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00008071,MFCD00008071
InChI Key DELJOESCKJGFML-DUXPYHPUSA-N
Synonym 3-aminocrotononitrile, 2z-3-aminobut-2-enenitrile, 3-amino-2-butenenitrile, z-3-aminobut-2-enenitrile, beta-aminocrotononitrile, 3-amino-2-butenonitrile, 2-amino-1-propenecarbonitrile, 3-amino-crotononitrile, beta-amino-crotononitrile, 2z-3-amino-2-butenenitrile
PubChem CID 5325263
IUPAC Name (Z)-3-aminobut-2-enenitrile
SMILES C\C(N)=C/C#N

Specifications

Melting Point ∼70°C to 75°C
Boiling Point 120°C (4 mmHg)
Flash Point 154°C (309°F)
Quantity 500 g
Beilstein 1719815
Solubility Information Very soluble in water,soluble in ethanol.
Formula Weight 82.11
Percent Purity 96%
Chemical Name or Material 3-Aminocrotononitrile, (E)+(Z)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.