missing translation for 'onlineSavingsMsg'
Learn More

3-Aminoisoxazole-4-carbonitrile, TRC

Product Code. 30330340
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity unitSize
30330340 50 mg 50mg
1 options
This item is not returnable. View return policy

Product Code. 30330340

Brand: Toronto Research Chemicals TRCA63607050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Aminoisoxazole-4-carbonitrile
CAS 258518-65-9
Molecular Formula C4 H3 N3 O
Quantity 50 mg
InChI Formula InChI=1S/C4H3N3O/c5-1-3-2-8-7-4(3)6/h2H,(H2,6,7)
SMILES N=C1NOC=C1C#N
IUPAC Name 3-imino-1,2-oxazole-4-carbonitrile
Molecular Weight (g/mol) 109.086
Formula Weight 109.028
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.