missing translation for 'onlineSavingsMsg'
Learn More

3-Aminoisoxazole, 95%

Product Code. 10748045 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10748045 5 mL Glass bottle 5mL
10386314 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10748045 Supplier Thermo Scientific Acros Supplier No. 133530050
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1750-42-1 | C3H4N2O | 84.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1750-42-1
Molecular Formula C3H4N2O
Molecular Weight (g/mol) 84.08
MDL Number MFCD00038814
InChI Key RHFWLPWDOYJEAL-UHFFFAOYSA-N
Synonym 3-aminoisoxazole, isoxazol-3-amine, 3-isoxazolamine, 3-amino isoxazole, isoxazolamine, 3-amineisoxazole, aminoisoxazole, 3-aminoisoxazol, 3-amino-1,2-oxazole, amino-isoxazole
PubChem CID 1810216
IUPAC Name 1,2-oxazol-3-amine
SMILES C1=CON=C1N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 1.3800g/mL
Boiling Point 226°C to 228°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5100 to 1.5120 (20°C, 589nm)
Quantity 5 mL
Specific Gravity 1.38
Formula Weight 84.08
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Aminoisoxazole
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminoisoxazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.