missing translation for 'onlineSavingsMsg'
Learn More

3-Aminomethyl-1-Boc-piperidine, 97%

Product Code. 15479607
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15479607 1 g 1g
15489607 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15479607 Supplier Thermo Scientific Alfa Aesar Supplier No. H52832.03
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 162167-97-7 | C11H22N2O2 | 214.309 g/mol

3-Aminomethyl-1-Boc-piperidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminomethyl-1-Boc-piperidine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162167-97-7
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD01317792
InChI Key WPWXYQIMXTUMJB-UHFFFAOYSA-N
Synonym tert-butyl 3-aminomethyl piperidine-1-carboxylate, 1-boc-3-aminomethylpiperidine, 1-boc-3-aminomethyl piperidine, 3-aminomethyl-1-n-boc-piperidine, 1-boc-3-aminomethyl-piperidine, 3-aminomethyl-1-boc-piperidine, 3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, n-boc-3-aminomethylpiperidine, 1-n-boc-3-aminomethyl-piperidine
PubChem CID 2756483
IUPAC Name tert-butyl 3-(aminomethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)CN

Specifications

Quantity 1 g
Formula Weight 214.31
Percent Purity 97%
Chemical Name or Material 3-Aminomethyl-1-Boc-piperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.