missing translation for 'onlineSavingsMsg'
Learn More

3-(Aminomethyl)pyridine, 99+%, ACROS Organics™

Product Code. 10489390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
100mL; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10489390 100mL; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10489390 Supplier Thermo Scientific Acros Supplier No. 104081000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 3731-52-0
CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C6H8N2
Refractive Index 1.5500 to 1.5520 (20°C, 589nm)
MDL Number MFCD00006412
Quantity 100 mL
Synonym 3-aminomethyl pyridine, 3-pyridinemethanamine, 3-picolylamine, picolamine, 3-pyridylmethylamine, 3-pyridinylmethanamine, 3-pyridinemethylamine, 3-aminomethylpyridine, pyridin-3-yl methanamine, 3-pyridylmethanamine
InChI Key HDOUGSFASVGDCS-UHFFFAOYSA-N
SMILES C1=CC(=CN=C1)CN
IUPAC Name pyridin-3-ylmethanamine
Molecular Weight (g/mol) 108.144
PubChem CID 31018
Formula Weight 108.14
Percent Purity ≥99.0%
Physical Form Liquid
Chemical Name or Material 3-(Aminomethyl)pyridine, 99+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.