missing translation for 'onlineSavingsMsg'
Learn More

3-Aminooxetane, 95%

Product Code. 15214874 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15214874 250 mg 250mg
15224874 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15214874 Supplier Thermo Scientific Acros Supplier No. 451212500
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 21635-88-1 | C3H7NO | 73.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21635-88-1
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.09
InChI Key OJEOJUQOECNDND-UHFFFAOYSA-N
Synonym 3-oxetanamine, 3-aminooxetane, oxetan-3-ylamine, 3-oxetanamin, 3-amino-oxetane, oxetane-3-amine, oxetan-3-yl amine, pubchem20414, acmc-1cgam
PubChem CID 9833923
IUPAC Name oxetan-3-amine
SMILES C1C(CO1)N

Specifications

Boiling Point 99°C
Flash Point 24°C
Assay Percent Range 94% min. (GC)
Quantity 250 mg
Formula Weight 73.09
Percent Purity 95%
Chemical Name or Material 3-Aminooxetane
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Aminooxetane, 95%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H301-Toxic if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.