missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophenylboronic acid hydrochloride, 98%

Product Code. 10537410 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10537410 1 g 1g
10027909 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10537410 Supplier Thermo Scientific Acros Supplier No. 435530010
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 85006-23-1 | C6H9BClNO2 | 173.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85006-23-1
Molecular Formula C6H9BClNO2
Molecular Weight (g/mol) 173.40
MDL Number MFCD00191748
InChI Key QBMHZZHJIBUPOX-UHFFFAOYSA-N
Synonym 3-aminophenyl boronic acid hydrochloride, 3-aminophenylboronic acid hydrochloride, 3-aminophenylboronic acid hcl, m-aminophenylboronic acid hydrochloride, 3-aminophenylboronic acid, hcl, 3-aminobenzeneboronic acid hydrochloride, boronic acid, 3-aminophenyl-, hydrochloride, pubchem1751, pubchem20396, timtec-bb sbb003816
PubChem CID 2763241
IUPAC Name (3-aminophenyl)boronic acid;hydrochloride
SMILES Cl.NC1=CC=CC(=C1)B(O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White to Gray
Elemental Analysis 40.52 to 42.60% C, 7.88 to 8.28% N
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 173.41
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminophenylboronic acid hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminophenylboronic acid hydrochloride, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.