missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophthalhydrazide, 98%, pure

Product Code. 11995651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11995651 100 g Glass bottle 100g
10480364 1 g Glass bottle 1g
10478120 5 g Glass bottle 5g
10511981 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11995651 Supplier Thermo Scientific Acros Supplier No. 153851000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3-Aminophthalhydrazide, 98%, pure, Quantity: 25g, Packaging: Glass bottle, Blue-Purple to Green, Grade: Pure, Melting Point: 319.0 deg.C to 320.0 deg.C, Molecular Weight: 177.16, Percent Purity: 98%, Assay Percent Range: 98%, Beilstein: 25,IV,4192, CAS: 521-31-3, CAS Max %: 100.0 | CAS: 521-31-3 | C8H7N3O2 | 177.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 521-31-3
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.16
MDL Number MFCD00006890
InChI Key HWYHZTIRURJOHG-UHFFFAOYSA-N
Synonym luminol, 3-aminophthalhydrazide, 3-aminophthalic hydrazide, 5-amino-2,3-dihydro-1,4-phthalazinedione, 1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalic acid hydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, unii-5exp385q4f, ccris 5962, 3-aminophthalylhydrazide
PubChem CID 10638
IUPAC Name 5-amino-2,3-dihydrophthalazine-1,4-dione
SMILES C1=CC2=C(C(=C1)N)C(=O)NNC2=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 319.0°C to 320.0°C
Color Blue-Purple to Green
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Beilstein 25,IV,4192
Merck Index 15,566
Solubility Information (2% in 1 N NaOH) Clear yellow to yellow-green
Formula Weight 177.16
Percent Purity 98%
Grade Pure
Physical Form Powder
Chemical Name or Material 3-Aminophthalhydrazide, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.