missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropionitrile, 98%, stabilized

Product Code. 11925621 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11925621

Brand: Thermo Scientific Acros 351750250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 151-18-8 | C3H6N2 | 70.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-18-8
Molecular Formula C3H6N2
Molecular Weight (g/mol) 70.10
MDL Number MFCD00014820
InChI Key AGSPXMVUFBBBMO-UHFFFAOYSA-N
Synonym 3-aminopropionitrile, 2-cyanoethylamine, aminopropionitrile, beta-aminopropionitrile, bapn, 3-aminopropiononitrile, beta-cyanoethylamine, propanenitrile, 3-amino, beta-alaninenitrile, propionitrile, 3-amino
PubChem CID 1647
ChEBI CHEBI:27413
IUPAC Name 3-aminopropanenitrile
SMILES NCCC#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9584g/mL
Boiling Point 185.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4365 to 1.4395
Quantity 25 g
Merck Index 15,465
Specific Gravity 0.9584
Solubility Information Solubility in water: miscible.
Stabilizer potassium carbonate
Formula Weight 70.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Aminopropionitrile, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Aminopropionitrile, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.