missing translation for 'onlineSavingsMsg'
Learn More

(3-Aminopropyl)trimethoxysilane, 97%

Product Code. 11403654
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403654 500 g 500g
11493644 100 g 100g
11413654 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403654 Supplier Thermo Scientific Alfa Aesar Supplier No. A11284.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13822-56-5 | C6H17NO3Si | 179.291 g/mol

(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).

Solubility
Miscible with water.

Notes
Moisture sensitive. Incompatible withstrong acids, strong oxidizing agents, water, bases, alcohols and peroxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13822-56-5
Molecular Formula C6H17NO3Si
Molecular Weight (g/mol) 179.291
MDL Number MFCD00008206
InChI Key SJECZPVISLOESU-UHFFFAOYSA-N
Synonym 3-aminopropyltrimethoxysilane, 3-aminopropyl trimethoxysilane, 3-trimethoxysilyl-1-propanamine, 3-trimethoxysilyl propan-1-amine, 3-trimethoxysilyl propylamine, 1-propanamine, 3-trimethoxysilyl, silane sc 3900, n-trimethoxysilylpropyl amine, propylamine, 3-trimethoxysilyl, unii-7mry0c9sb6
PubChem CID 83756
IUPAC Name 3-trimethoxysilylpropan-1-amine
SMILES CO[Si](CCCN)(OC)OC

Specifications

Density 1.02
Boiling Point 194°C
Flash Point 90°C (194°F)
Odor Amine-like
Linear Formula H2N(CH2)3Si(OCH3)3
Refractive Index 1.424
Quantity 500 g
UN Number UN2735
Beilstein 2038118
Sensitivity Air and moisture sensitive
Solubility Information Miscible with water.
Formula Weight 179.29
Percent Purity 97%
Chemical Name or Material (3-Aminopropyl)trimethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.