missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyrazine-2-carboxylic acid, 98%

Product Code. 11440128
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440128 5 g 5g
11430128 1 g 1g
11450128 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440128 Supplier Thermo Scientific Alfa Aesar Supplier No. B23707.06
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5424-01-1 | C5H5N3O2 | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1-1-5424
MDL Number MFCD00006141

Specifications

Melting Point 204°C to 206°C (decomposition)
Molecular Formula C5H5N3O2
Quantity 5 g
Beilstein 124835
Synonym 3-amino-2-pyrazinecarboxylic acid, pyrazinecarboxylic acid, 3-amino, 2-amino-3-pyrazinecarboxylic acid, 3-aminopiperazine-2-carboxylic acid, 2-aminopyrazine-3-carboxylic acid, 2-pyrazinecarboxylic acid, 3-amino, 3-aminopyrazine-2-carboxyliacid, zlchem 459, pubchem8544, 3-aminopyrazinoic acid
InChI Key ZAGZIOYVEIDDJA-UHFFFAOYSA-N
SMILES C1=CN=C(C(=N1)C(=O)O)N
IUPAC Name 3-aminopyrazine-2-carboxylic acid
Molecular Weight (g/mol) 139.11
PubChem CID 72656
Formula Weight 139.11
Percent Purity ≥98%
Chemical Name or Material 3-Aminopyrazine-2-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminopyrazine-2-carboxylic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 3-Aminopyrazine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.