missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyrazine-2-carboxylic acid, 99+%

Product Code. 10436900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10436900 5 g Glass bottle 5g
10375970 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10436900 Supplier Thermo Scientific Acros Supplier No. 104500050
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5424-01-1 | C5H5N3O2 | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1-1-5424
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.11
MDL Number MFCD00006141
InChI Key ZAGZIOYVEIDDJA-UHFFFAOYSA-N
Synonym 3-amino-2-pyrazinecarboxylic acid, pyrazinecarboxylic acid, 3-amino, 2-amino-3-pyrazinecarboxylic acid, 3-aminopiperazine-2-carboxylic acid, 2-aminopyrazine-3-carboxylic acid, 2-pyrazinecarboxylic acid, 3-amino, 3-aminopyrazine-2-carboxyliacid, zlchem 459, pubchem8544, 3-aminopyrazinoic acid
PubChem CID 72656
IUPAC Name 3-aminopyrazine-2-carboxylic acid
SMILES C1=CN=C(C(=N1)C(=O)O)N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 201°C to 205°C
Color Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
Formula Weight 139.11
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminopyrazine-2-carboxylic acid, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminopyrazine-2-carboxylic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.