missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyrazole, 98%

Product Code. 10762021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10762021 2.5 g Glass bottle 2.5g
10361384 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10762021 Supplier Thermo Scientific Acros Supplier No. 171030025
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1820-80-0 | C3H5N3 | 83.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1820-80-0
Molecular Formula C3H5N3
Molecular Weight (g/mol) 83.09
MDL Number MFCD00005236
InChI Key JVVRJMXHNUAPHW-UHFFFAOYSA-N
Synonym 3-aminopyrazole, 1h-pyrazol-3-amine, 1h-pyrazole-5-amine, 5-aminopyrazole, 1h-pyrazol-3-ylamine, 3-amino-1h-pyrazole, 3-aminopyrozole, 5-amino-1h-pyrazole, aminopyrazole, 3-pyrazolamine
PubChem CID 74561
IUPAC Name 1H-pyrazol-5-amine
SMILES C1=C(NN=C1)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 33.0°C to 39.0°C
Color White to Yellow
Boiling Point 218.0°C (122.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 2.5 g
Beilstein 24, 14
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
Formula Weight 83.09
Percent Purity 98%
Physical Form Oily Liquid or Low Melting Crystalline Solid
Chemical Name or Material 3-Aminopyrazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Aminopyrazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.