missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyridazine, 95%, Thermo Scientific™

Product Code. 15750889 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15750889 250 mg 250mg
15740889 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15750889 Supplier Thermo Scientific Acros Supplier No. 454172500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5469-70-5
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.1
InChI Key LETVJWLLIMJADE-UHFFFAOYSA-N
Synonym 3-aminopyridazine, 3-pyridazinamine, aminopyridazine, pyridazin-3-ylamine, 3-amino-1,2-diazine, pyridazine-3-ylamine, pyridazineamine, pyridazyl amine, imino-pyridazine, 3-pyridazinamin
PubChem CID 230373
IUPAC Name pyridazin-3-amine
SMILES C1=CC(=NN=C1)N

Specifications

Melting Point 168.0°C to 172.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 95.1
Percent Purity 95%
Chemical Name or Material 3-Aminopyridazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.