missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyridazine, 97%

Product Code. p-7056104
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15402259 1 g 1g
15412259 5 g 5g
15422259 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15402259 Supplier Thermo Scientific Alfa Aesar Supplier No. H64859.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5469-70-5 | C4H5N3 | 95.105 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5469-70-5
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.105
MDL Number MFCD01529869
InChI Key LETVJWLLIMJADE-UHFFFAOYSA-N
Synonym 3-aminopyridazine, 3-pyridazinamine, aminopyridazine, pyridazin-3-ylamine, 3-amino-1,2-diazine, pyridazine-3-ylamine, pyridazineamine, pyridazyl amine, imino-pyridazine, 3-pyridazinamin
PubChem CID 230373
IUPAC Name pyridazin-3-amine
SMILES C1=CC(=NN=C1)N

Specifications

Melting Point 168°C to 172°C
Color White to Yellow
Quantity 1 g
Beilstein 107024
Formula Weight 95.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Aminopyridazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.