missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyridine, TRC

Product Code. 30010298
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010298 50 g 50g
30011483 10 g 10g
30009926 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010298 Supplier Toronto Research Chemicals Supplier No. TRCA62901550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-08-8
Molecular Formula C5 H6 N2
Molecular Weight (g/mol) 94.11
Synonym Pyridine, 3-amino- (8CI), 3-Aminopyridine, 3-Pyridinylamine, 3-Pyridylamine, NSC 15040, m-Aminopyridine, β-Aminopyridine
IUPAC Name pyridin-3-amine
SMILES Nc1cccnc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C5H6N2/c6-5-2-1-3-7-4-5/h1-4H,6H2
Formula Weight 94.0531
Chemical Name or Material 3-Aminopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.