missing translation for 'onlineSavingsMsg'
Learn More

3-Aminotetrahydro-1H-1lambda∼6∼-thiophene-1,1-dione hydrochloride , Tech., Thermo Scientific™

Product Code. p-3485445
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10739183 250 mg Amber Glass Bottle 250mg
10095112 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10739183 Supplier Thermo Scientific Maybridge Supplier No. RJC02399CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 51642-03-6
Molecular Formula C4H10ClNO2S
Molecular Weight (g/mol) 171.64
MDL Number MFCD00456584
InChI Key MGZQMSFXPSKBDY-UHFFFAOYNA-N
Synonym 3-aminotetrahydrothiophene 1,1-dioxide hydrochloride, 1,1-dioxidotetrahydrothien-3-ylamine hydrochloride, tetrahydro-3-thiophenamine 1,1-dioxide hydrochloride, 1,1-dioxo-tetrahydrothiophen-3-ylamine, hcl, 3-amino-1??-thiolane-1,1-dione hydrochloride, 1,1-dioxidotetrahydro-3-thienyl amine hydrochloride, 3-aminotetrahydrothiopene 1,1-dioxide hydrochloride, tetrahydrothiophen-3-amine 1,1-dioxide hydrochloride, 3-thiophenamine, tetrahydro-, 1,1-dioxide, hydrochloride, 1,1-dioxothiolan-3-amine hydrochloride
PubChem CID 2795201
SMILES [H+].[Cl-].NC1CCS(=O)(=O)C1

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name hydrogen 3-amino-1λ⁶-thiolane-1,1-dione chloride
Formula Weight 171.65
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-Aminotetrahydrothiopene 1, 1-dioxide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.