missing translation for 'onlineSavingsMsg'
Learn More

3-arylisoquinolinamine derivative, MedChemExpress

Product Code. 16478038
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478038 10 mg 10mg
16473957 50 mg 50mg
16427816 100 mg 100mg
16410508 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16478038 Supplier Medchem Express Supplier No. HY32364
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 3-arylisoquinolinamine derivative is a 3-arylisoquinolinamine derivative with antitumor activity.

3-arylisoquinolinamine derivative is a 3-arylisoquinolinamine derivative with antitumor activity.

Specifications

Chemical Name or Material 3-arylisoquinolinamine derivative
CAS 1029008-71-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H19N3O
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (170.44 mM)
SMILES NC1=NC(C2=CC=CC(OC)=C2)=CC(C=C3)=C1C=C3N(C)C
Molecular Weight (g/mol) 293.36
Formula Weight 293.36
Percent Purity 99.1%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.