missing translation for 'onlineSavingsMsg'
Learn More

3-Benzoylpropionic acid, 98+%

Product Code. 11497967
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497967 100 g 100g
11487967 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11497967 Supplier Thermo Scientific Alfa Aesar Supplier No. A16636.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2051-95-8 | C10H10O3 | 178.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-95-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00002792
InChI Key KMQLIDDEQAJAGJ-UHFFFAOYSA-N
Synonym 3-benzoylpropionic acid, 4-oxo-4-phenylbutyric acid, 3-benzoylpropanoic acid, beta-benzoylpropionic acid, benzoylpropionic acid, propanoic acid, 3-benzoyl, benzenebutanoic acid, .gamma.-oxo, 3-benzoylpropionicacid, propionic acid, 3-benzoyl, 4-phenyl-4-oxobutyric acid
PubChem CID 72871
ChEBI CHEBI:64437
IUPAC Name 4-oxo-4-phenylbutanoic acid
SMILES OC(=O)CCC(=O)C1=CC=CC=C1

Specifications

Melting Point 114°C to 120°C
Quantity 100 g
Beilstein 639757
Formula Weight 178.19
Percent Purity ≥98%
Chemical Name or Material 3-Benzoylpropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.