missing translation for 'onlineSavingsMsg'
Learn More

3-Benzylidene-2,4-pentanedione, 97%

Product Code. 11340134
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11340134 5 g 5g
11350134 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11340134 Supplier Thermo Scientific Alfa Aesar Supplier No. L00623.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4335-90-4 | C12H12O2 | 188.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4335-90-4
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.23
MDL Number MFCD00014990
InChI Key NYRGMNMVISROGJ-UHFFFAOYSA-N
Synonym 3-benzylidene-2,4-pentanedione, benzalacetylacetone, benzylidene acetylacetone, 3-benzylideneacetylacetone, 2,4-pentanedione, 3-phenylmethylene, 1,1-diacetyl-2-phenylethylene, 2,2-diacetylvinyl benzene, 2-acetyl-1-phenyl but-1-en-3-one, 2,4-pentanedione, 3-benzylidene, 3-benzylidene acetylacetone
PubChem CID 95782
IUPAC Name 3-benzylidenepentane-2,4-dione
SMILES CC(=O)C(=CC1=CC=CC=C1)C(C)=O

Spécification

Density 1.093
Boiling Point 185°C to 188°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 972849
Formula Weight 188.23
Percent Purity 97%
Chemical Name or Material 3-Benzylidene-2,4-pentanedione

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.