missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxy-4-methoxybenzaldehyde, TRC

Product Code. 30338645
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30338645 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338645 Supplier Toronto Research Chemicals Supplier No. TRCB28598010G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Benzyloxy-4-methoxybenzaldehyde
CAS 6346-05-0
Molecular Formula C15 H14 O3
Quantity 10 g
Synonym Benzaldehyde, 4-methoxy-3-(phenylmethoxy)-, Benzaldehyde, 3-(benzyloxy)-4-methoxy- (6CI,8CI), 4-Methoxy-3-(phenylmethoxy)benzaldehyde, 3-(Benzyloxy)-4-methoxybenzaldehyde, 4-Methoxy-3-benzyloxybenzaldehyde, Benzylisovanillin, Isovanillin benzyl ether, NSC 196547, NSC 43750, O-Benzylisovanillin, 3-Benzyloxy-4-methoxybenzaldehyde
InChI Formula InChI=1S/C15H14O3/c1-17-14-8-7-13(10-16)9-15(14)18-11-12-5-3-2-4-6-12/h2-10H,11H2,1H3
SMILES COc1ccc(C=O)cc1OCc2ccccc2
IUPAC Name 4-methoxy-3-phenylmethoxybenzaldehyde
Molecular Weight (g/mol) 242.27
Formula Weight 242.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.