missing translation for 'onlineSavingsMsg'
Learn More

3-(Benzyloxycarbonylamino)azetidine, 97%

Product Code. 15449607
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15449607 1 g 1g
15469607 250 mg 250mg
15459607 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15449607 Supplier Thermo Scientific Alfa Aesar Supplier No. H52831.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 914348-04-2 | C11H14N2O2 | 206.245 g/mol

3-(Benzyloxycarbonylamino)azetidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Benzyloxycarbonylamino)azetidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 914348-04-2
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.245
MDL Number MFCD07368913
InChI Key UWOOBFRZDNDUQB-UHFFFAOYSA-N
Synonym benzyl azetidin-3-ylcarbamate, 3-cbz-amino azetidine, 3-cbz-amino-azetidine, benzyl n-azetidin-3-yl carbamate, carbamic acid, n-3-azetidinyl-, phenylmethyl ester, azetidin-3-yl-carbamic acid benzyl ester, 3-benzyloxycarbonylamino azetidine, pubchem10159, 3-n-cbz-amino azetidine, benzyl azetidine-3-ylcarbamate
PubChem CID 17750122
IUPAC Name benzyl N-(azetidin-3-yl)carbamate
SMILES C1C(CN1)NC(=O)OCC2=CC=CC=C2

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 3-(Benzyloxycarbonylamino)azetidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.