missing translation for 'onlineSavingsMsg'
Learn More

3-(Benzyloxycarbonylamino)azetidine, 97%

Product Code. 15459607
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459607 5 g 5g
15469607 250 mg 250mg
15449607 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15459607 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor H52831.06
 Restricted Item
Estimated Shipment: 21-12-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 914348-04-2 | C11H14N2O2 | 206.245 g/mol

3-(Benzyloxycarbonylamino)azetidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Benzyloxycarbonylamino)azetidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 914348-04-2
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.245
MDL Number MFCD07368913
InChI Key UWOOBFRZDNDUQB-UHFFFAOYSA-N
Synonym benzyl azetidin-3-ylcarbamate, 3-cbz-amino azetidine, 3-cbz-amino-azetidine, benzyl n-azetidin-3-yl carbamate, carbamic acid, n-3-azetidinyl-, phenylmethyl ester, azetidin-3-yl-carbamic acid benzyl ester, 3-benzyloxycarbonylamino azetidine, pubchem10159, 3-n-cbz-amino azetidine, benzyl azetidine-3-ylcarbamate
PubChem CID 17750122
IUPAC Name benzyl N-(azetidin-3-yl)carbamate
SMILES C1C(CN1)NC(=O)OCC2=CC=CC=C2

Especificaciones

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 3-(Benzyloxycarbonylamino)azetidine

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.